Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD366 (TIM-3) Rat anti-Mouse, BV480, Clone: RMT3-23, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 568829
Description
CD366 (TIM-3) Monoclonal antibody specifically detects CD366 (TIM-3) in Mouse samples. It is validated for Flow cytometry
Specifications
CD366 (TIM-3) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
171285 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2a κ |
Flow Cytometry | |
RMT3-23 | |
Brilliant Violet 480 | |
Cd366; TIM-3; Tim3; TIMD-3; Timd3; HAVcr-2; Havcr2 | |
Mouse TIM-3 Recombinant Protein | |
0.1 mg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction