Learn More
CD368 (Clec6) Monoclonal Antibody (9B9), eFluor™ 660, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$186.00 - $453.00
Specifications
Antigen | CD368 (Clec6) |
---|---|
Clone | 9B9 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-911-341
![]() |
Invitrogen™
50911341 |
25 Tests |
Each for $186.00
|
|
|||||
50-911-342
![]() ![]() |
Invitrogen™
50911342 |
100 Tests |
Each for $453.00
|
|
|||||
Description
This 9B9 MAb reacts with human CD368, also known as Clec6 or ClecsF8 or Clec4D. In peripheral blood, this C-type lectin is expressed highly on granulocytes and CD14+CD16- monocytes. Some dendritic cell populations also exhibit low levels of CD368 expression. Studies have demonstrated that this receptor undergoes endocytosis due to its C-type lectin-like domain. CD368 functions as a phagocytic receptor and is involved in proinflammatory cytokine production in cooperation with Toll-like receptors. Finally, CD368 also participates in cellular activation via signaling through Syk kinase.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.Specifications
CD368 (Clec6) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
CD368; Clec4d; CLEC6; CLEC-6; CLECSF8; CLECSF8Mpcl; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 8; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 8; C-type lectin domain family 4 member D; C-type lectin domain family 4, member d; C-type lectin receptor; C-type lectin superfamily member 8; C-type lectin, superfamily member 8; C-type lectin-like receptor 6; macrophage C-type lectin; macrophage-restricted C-type lectin; Mcl; MGC40078; Mpcl; superfamilymember 8 | |
CLEC4D | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
CLEC4D |
9B9 | |
Flow Cytometry | |
eFluor 660 | |
Mouse | |
Human | |
Q8WXI8 | |
338339 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.