Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD37 Antibody (2B8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128869
Description
CD37 Monoclonal specifically detects CD37 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CD37 | |
Monoclonal | |
Unconjugated | |
Ascites with 0.03% Sodium Azide | |
CD37 | |
Purified recombinant fragment of CD37 expressed in E. Coli. | |
Unpurified | |
RUO | |
951 | |
Human, Mouse | |
Ascites |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
2B8 | |
Flow Cytometry, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin | |
CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37 | |
Mouse | |
31.7 kDa | |
0.1 mL | |
Primary | |
Specific for CD37. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction