Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD37 Antibody (WR17), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281048JF549
Description
CD37 Monoclonal specifically detects CD37 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CD37 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
CD37 | |
Chronic lymphocytic leukaemia cells. | |
0.1 mL | |
Cellular Markers, Immunology | |
951 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
WR17 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37 | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Recognises human CD37. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction