Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD37 Chimeric Recombinant Rabbit Monoclonal Antibody (G28-1)

Rabbit Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA548355

Catalog No. PIMA548355


Only null left
Add to Cart

Description

Description

Specificity: This antibody recognises CD37, which is a cell surface protein, present mostly on mature B cells and B cell malignancies.

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It may play a role in T-cell-B-cell interactions. Alternate splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD37 Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P11049
CD37
This antibody was raised by immunizing mice with CD37.
200 μg
Primary
Human
Antibody
IgG κ
Flow Cytometry, Functional Assay, Immunocytochemistry
G28-1
Unconjugated
CD37
CD37; CD37 antigen; CD37 molecule; cell differentiation antigen 37; GP52-40; Leukocyte antigen CD37; leukocyte surface antigen CD37; tetraspanin-26; Tspan26; Tspan-26
Rabbit
Protein A
RUO
951
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.