Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD37 Monoclonal Antibody (MB-1), APC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 501123047
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-3047 100 Tests
50-112-3046 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-3047 Supplier Invitrogen™ Supplier No. 17037942
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This MB-1 monoclonal antibody recognizes human CD37. This tetraspanin is expressed on mature B cells, but not on pro-B or plasma cells. CD37 is also weakly expressed on T cells, monocytes, neutrophils, dendritic cells, and macrophages. This cell surface glycoprotein is not detected on NK cells, platelets, or erythrocytes. CD37 is highly expressed on all forms of B cell non-Hodgkin lymphoma, chronic lymphocyte leukemia, and other leukemias. CD37 reportedly associates with MHC class II, CD19, CD21, and other members of the tetraspanin family. CD37 has been implicated in signal transduction, B cell activation, and T cell proliferation. CD37 has been used as a therapeutic target for treating B cell malignancies. Applications Reported: This MB-1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MB-1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It may play a role in T-cell-B-cell interactions. Alternate splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD37
Applications Flow Cytometry
Classification Monoclonal
Clone MB-1
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CD37
Gene Accession No. P11049
Gene Alias CD37; CD37 antigen; CD37 molecule; cell differentiation antigen 37; GP52-40; Leukocyte antigen CD37; leukocyte surface antigen CD37; tetraspanin-26; Tspan26; Tspan-26
Gene Symbols CD37
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 951
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.