Learn More
CD37 Mouse anti-Human, Brilliant Violet 650, Clone: M-B371, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 742400
Description
The M-B371 monoclonal antibody specifically recognizes CD37. CD37 is a 40-52 kDa type II integral membrane glycoprotein present on B cells from the pre-B-cell stage but not on plasma cells. CD37 is also expressed on activated, proliferating cells of germinal centers. There is low expression of CD37 on some T and myeloid cells. The function of CD37 has not yet been clearly identified.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
Specifications
CD37 | |
M-B371 | |
Brilliant Violet 650 | |
GP52-40, Tetraspanin-26, TSPAN26, Tspan-26 | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.