Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD37 Rabbit anti-Human, Clone: IPO-24 (ZC37-24), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP3120740.025MG
Description
CD37 Monoclonal antibody specifically detects CD37 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
CD37 | |
Monoclonal | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37 | |
This antibody was raised by immunizing mice with spleen cells of a patient with hairy cell leukemia. | |
0.025 mg | |
Cellular Markers, Immunology | |
951 | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
IPO-24 (ZC37-24) | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction