Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD37, Rabbit anti-Human, Clone: WR17, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | CD37 |
---|---|
Clone | WR17 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence |
Description
CD37 Monoclonal specifically detects CD37 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CD37 | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cellular Markers, Immunology | |
PBS | |
951 | |
Chronic lymphocytic leukaemia cells. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
WR17 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37 | |
CD37 | |
IgG κ | |
Protein A purified | |
Recognises human CD37. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title