Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD37 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 210441AP150UL

Description
CD37 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It may play a role in T-cell-B-cell interactions. Alternate splicing results in multiple transcript variants encoding different isoforms.Specifications
CD37 | |
Polyclonal | |
Unconjugated | |
CD37 | |
CD37, CD37 molecule, GP52 40, Leukocyte antigen CD37, Tetraspanin 26, Tspan 26, TSPAN26 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12493, 29185, 951 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P11049, P31053, Q61470 | |
Cd37 | |
CD37 Fusion Protein Ag15384 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction