Learn More
CD370 (CLEC9A/DNGR1) Monoclonal Antibody (1F6), Functional Grade, In Vivo GOLD™, Leinco Technologies™

Mouse Monoclonal Antibody
Supplier: Leinco Technologies Inc I2001100MG


Description
Specificity: This clone 1F6 monoclonal antibody recognizes a direct epitope on mouse DNGR1/CLEC9A for CD8 positive dendritic cells, which can be exploited for tumor therapy, whilst also recognizing necrotic cells. This antibody can recognize both the short and long isoform of DNGR-1, which varies in a segment of the stalk region. Endotoxin Level: <1.0 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
CD370 (CLEC9A/DNGR1) Monoclonal antibody specifically detects CD370 (CLEC9A/DNGR1) in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry, Western BlotSpecifications
CD370 (CLEC9A/DNGR1) | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
Q8BRU4 | |
Mouse | |
Protein A/G | |
RUO | |
232414 | |
4°C short term, -80°C long term | |
Liquid |
Flow Cytometry, Functional Assay, Immunocytochemistry, Western Blot | |
1F6 | |
Functional Grade | |
CLEC9A | |
9830005G06Rik; CD370; Clec9a; C-type lectin domain family 9 member A; C-type lectin domain family 9, member A; Dendritic cell natural killer lectin group receptor 1; DNGR1; DNGR-1; HEEE9341; RGD1562513; UNQ9341; UNQ9341/PRO34046 | |
RBL-2H3 cells expressing mouse DNGR-1 fused to an HA epitope | |
100 mg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.