Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD370 (CLEC9A/DNGR1) Monoclonal Antibody (1F6), Functional Grade, In Vivo PLATINUM™, Leinco Technologies™
SDP

Catalog No. 502544147
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4147 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4147 Supplier Leinco Technologies Inc Supplier No. I2020100MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specificity: This clone 1F6 monoclonal antibody recognizes a direct epitope on mouse DNGR1/CLEC9A for CD8 positive dendritic cells, which can be exploited for tumor therapy, whilst also recognizing necrotic cells. This antibody can recognize both the short and long isoform of DNGR-1, which varies in a segment of the stalk region. Endotoxin Level: ≤ 0.5 EU/mg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco Purified Functional PLATINUM™ antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.

CD370 (CLEC9A/DNGR1) Monoclonal antibody specifically detects CD370 (CLEC9A/DNGR1) in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry, Western Blot

Specifications

Antigen CD370 (CLEC9A/DNGR1)
Applications Flow Cytometry, Functional Assay, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1F6
Concentration ≥5.0 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2-7.4
Gene CLEC9A
Gene Accession No. Q8BRU4
Gene Alias 9830005G06Rik; CD370; Clec9a; C-type lectin domain family 9 member A; C-type lectin domain family 9, member A; Dendritic cell natural killer lectin group receptor 1; DNGR1; DNGR-1; HEEE9341; RGD1562513; UNQ9341; UNQ9341/PRO34046
Host Species Mouse
Immunogen RBL-2H3 cells expressing mouse DNGR-1 fused to an HA epitope
Purification Method Protein A/G
Quantity 100 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232414
Target Species Mouse
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.