Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD370 (Clec9A) Rat anti-Mouse, BB515, Clone: 10B4, BD Horizon™
Rat Monoclonal Antibody
Specifications
| Antigen | CD370 (Clec9A) |
|---|---|
| Clone | 10B4 (also known as 24/04-10B4) |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BDB565320
|
BD Biosciences
565320 |
50 μg | N/A | N/A | N/A | ||||
Description
CD370 (Clec9A) Monoclonal antibody specifically detects CD370 (Clec9A) in Mouse samples. It is validated for Flow cytometry.
CD370 (Clec9A) Monoclonal antibody specifically detects CD370 (Clec9A) in Mouse samples. It is validated for Flow cytometry.Specifications
| CD370 (Clec9A) | |
| 0.2mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD370; CLC9A; Clec9a; DNGR-1; C-type lectin domain family 9 member A | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. |
| 10B4 (also known as 24/04-10B4) | |
| Flow Cytometry | |
| Brilliant Blue 515 (BB515) | |
| Rat | |
| Cell Biology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse Clec9A Peptide | |
| Primary |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title