Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD370 (Clec9A) Rat anti-Mouse, Brilliant Violet 786, Clone: 10B4 (also known as 24/04-10B4), BD Optibuild™

Catalog No. BDB744514 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744514 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744514 Supplier BD Biosciences Supplier No. 744514
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 10B4 monoclonal antibody specifically binds to mouse Clec9A. Mouse Clec9A (C-type lectin domain family member 9A) is also known as DNGR1 (Dendritic cell natural killer lectin group receptor 1). It is a type II membrane protein with a single extracellular C-type lectin domain. Clec9A is a dendritic cell subtype-restricted C-type lectin-like receptor. Clec9A is selectively expressed on plasmacytoid dendritic cells and CD8+ myeloid dendritic cells. Clec9A reportedly serves as a receptor for necrotic cells. It can mediate the cross-presentation of dead-cell associated antigens in a Syk-dependent manner.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD370 (Clec9A)
Classification Monoclonal
Clone 10B4 (also known as 24/04-10B4)
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD370, CLC9A, Clec9a, DNGR-1, C-type lectin domain family 9 member A
Host Species Rat
Immunogen Mouse Clec9A Peptide
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.