Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD370 (Clec9A) Rat anti-Mouse, Oligo, Clone: 10B4 (also known as 24/04-10B4), BD™ AbSeq ™

Catalog No. BDB940340 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB940340 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB940340 Supplier BD Biosciences Supplier No. 940340
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD370 (Clec9A) Monoclonal antibody specifically detects CD370 (Clec9A) in Mouse samples. It is validated for Single Cell 3' Sequencing
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD370 (Clec9A)
Applications Single Cell 3' Sequencing
Classification Monoclonal
Clone Clone 10B4 (also known as 24/04-10B4)
Conjugate Oligo
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias CD370; CLC9A; Clec9a; DNGR-1; C-type lectin domain family 9 member A
Host Species Rat
Immunogen Mouse Clec9A Peptide
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.