Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD371 (Clec12A) Mouse anti-Human, BUV395, Clone: 50C1, BD Optibuild™

Catalog No. BDB742935 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742935 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742935 Supplier BD Biosciences Supplier No. 742935
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 50C1 monoclonal antibody specifically binds to human CD371 which is also known as Clec12A (C-type lectin domain family 12 member A), C-type lectin-like molecule 1 (CLL-1), myeloid inhibitory C-type lectin-like receptor (MICL), or dendritic cell-associated lectin 2 (DCAL-2). It is expressed on a variety of cells including monocytes, macrophages, dendritic cells, and granulocytes and perhaps some NK cells. Clec12A is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. It is a 30 kDa type II transmembrane glycoprotein that has one single C-type lectin-like domain and one cytoplasmic immunoreceptor tyrosine-based inhibitory motif (ITIM). Clec12A has similarity with theβ-glucan receptor (Dectin-1) and LOX-1 with high N-glycosylation. There are at least five isoforms due to alternative transcript splicing. Signaling through Clec12A can induce internalization of Clec12A, dendritic cell maturation and the production of cytokines including IL-12. Clec12A may also serve as a negative regulator of activated leukocytes recruited to sites of inflammation

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD371 (Clec12A)
Classification Monoclonal
Clone 50C1
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD371, Clec12A, MICL, CLL-1, DCAL-2
Host Species Mouse
Immunogen Human CLEC12A Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.