Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD38 Antibody (HB7) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$280.00 - $596.50
Specifications
Antigen | CD38 |
---|---|
Clone | HB7 |
Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25264725
![]() |
Novus Biologicals
NBP2526470.025MG |
0.025 mg |
Each for $280.00
|
|
|||||
NBP25264702
![]() |
Novus Biologicals
NBP2526470.2MG |
0.2 mg |
Each for $596.50
|
|
|||||
Description
CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CD38 | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10 | |
CD38 | |
IgG | |
Protein A or G purified | |
This antibody binds to an epitope between amino acids 273-285 of human CD38, an approximately 45 kDa type II transmembrane protein, expressed on essentially all pre-B lymphocytes, plasma cells, and thymocytes. Also present on activated T lymphocytes, natural killer (NK) lymphocytes, myeloblasts, and erythroblasts. Bimodally expressed during B cell development, modulating from high in immature cells to low in intermediate ones and back to high on mature B cells. This antibody competes with clone AT13/5 (Ab00289) (Ellis 1995). |
HB7 | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Rabbit | |
Apoptosis, B Cell Development and Differentiation Markers, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Stem Cell Markers | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
952 | |
Viable cells (1 x 10E7) of the human cell line, BJAB (Human Burkitt lymphoma B cell line), which was were injected intraperitoneally into 8-week-old BALB/c mice. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title