Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD38 Antibody (OTI1C9), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270376G
Description
CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CD38 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD38 | |
Recombinant protein fragment corresponding to amino acids 43-300 of human CD38 (NP_001766) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1C9 | |
Western Blot, Immunohistochemistry | |
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10 | |
Mouse | |
Affinity Purified | |
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers | |
952.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction