Learn More
CD38 Mouse anti-Human, BV650, Clone: HB7 (also known as HB-7), BD Horizon™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD38 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HB7 (also known as HB-7) |
| Conjugate | Brilliant Violet 650 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | T10; ADP-ribosyl cyclase 1; Cyclic ADP-ribose hydrolase 1; OKT10 |
| Host Species | Mouse |
| Immunogen | Human BJAB B cell line |
| Purification Method | Affinity Chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.