Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD38 Mouse anti-Human, Clone: HB7 (APC-C750), Abnova™

Catalog No. 89033959 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-959 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-959 Supplier Abnova Corporation Supplier No. MAB13894
Add to Cart
Add to Cart
Request a Quote

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human CD38.

  • High specificity
  • High reactivity

Specifications

Antigen CD38
Applications Flow Cytometry
Classification Monoclonal
Clone HB7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human CD38.
Dilution Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Gene Accession No. P28907
Gene Alias T10
Gene Symbols CD38
Host Species Mouse
Immunogen BJAB human B cell line.
Purification Method Protein A/G purification
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 952
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.