Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD38 Mouse anti-Human, Mouse, Clone: 3C6G4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CD38 |
---|---|
Clone | 3C6G4 |
Concentration | 0.88 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6188
![]() ![]() |
Proteintech Group Inc
600061IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
CD38 (NAD+ glycohydrolase) is a type II transmembrane glycoprotein able to induce activation, proliferation and differentiation of mature lymphocytes and mediate apoptosis of myeloid and lymphoid progenitor cells. CD38 functions as a multi-catalytic ectoenzyme serving as ADP-ribosyl cyclase, cyclic ADP-ribose hydrolase and possibly NAD+ glycohydrolase or as a cell surface receptor. Antibodies to CD38 are useful in subtyping of lymphomas and leukemias, detection of plasma cells (i.e. identification of myelomas), and as a marker for activated B and T cells. CD38 participates in cell adhesion, signal transduction and calcium signaling. Further, CD38 is expressed at high levels in the pancreas, liver, kidney, maligt lymphoma and neuroblastoma. Diseases associated with CD38 dysfunction include chronic lymphocytic leukemia and Richter′s Syndrome.Specifications
CD38 | |
0.88 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ADP ribosyl cyclase 1, cADPr hydrolase 1, CD38, CD38 molecule, Cyclic ADP ribose hydrolase 1, T10 | |
CD38 | |
IgG1 | |
Protein G | |
Antibody |
3C6G4 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P28907, P56528 | |
12494, 952 | |
CD38 Fusion Protein Ag0487 | |
Primary | |
-20°C | |
CD38 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title