Learn More
CD38 Mouse anti-Human, PE, Clone: HB7, BD
Mouse Monoclonal Antibody
$380.00 - $608.50
Specifications
Antigen | CD38 |
---|---|
Clone | HB7 |
Concentration | 12.5μg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The CD38 antibody, clone HB7, is derived from hybridization of mouse P3-X63-Ag8.653 myeloma cells with spleen cells from BALB/c mice immunized with the BJAB cell line.
The CD38 antibody recognizes an integral membrane glycoprotein of 45 kilodaltons (kDa), with a protein core of 35 kDa. The CD38 antigen is also known as T10, ADP-ribosyl cyclase, and cyclic ADP ribose hydrolase 1.
Specifications
CD38 | |
12.5μg/mL | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
HB7 | |
Flow Cytometry | |
PE | |
RUO | |
Human BJAB B cell line | |
Primary |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.