Learn More
Invitrogen™ CD38 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589326
Description
Positive Samples: Raji; Cellular Location: Membrane, Single-pass type II membrane protein Immunogen sequence: VPRWRQQWSG PGTTKRFPET VLARCVKYTE IHPEMRHVDC QSVWDAFKGA FISKHPCNIT EEDYQPLMKL GTQTVPCNKI LLWSRIKDLA HQFTQVQRDM FTLEDTLLGY LADDLTWCGE FNTSKINYQS CPDWRKDCSN NPVSVFWKTV SRRFAEAACD VVHVMLNGSR SKIFDKNSTF GSVEVHNLQP EKVQTLEAWV IHGGREDSRD LCQDPTIKEL ESIISKRNIQ FSCKNIYRPD KFLQCVKNPE DSSCTSEI.
CD38 (NAD+ glycohydrolase) is a type II transmembrane glycoprotein able to induce activation, proliferation and differentiation of mature lymphocytes and mediate apoptosis of myeloid and lymphoid progenitor cells. CD38 functions as a multi-catalytic ectoenzyme serving as ADP-ribosyl cyclase, cyclic ADP-ribose hydrolase and possibly NAD+ glycohydrolase or as a cell surface receptor. Antibodies to CD38 are useful in subtyping of lymphomas and leukemias, detection of plasma cells (i.e. identification of myelomas), and as a marker for activated B and T cells. CD38 participates in cell adhesion, signal transduction and calcium signaling. Further, CD38 is expressed at high levels in the pancreas, liver, kidney, malignant lymphoma and neuroblastoma. Diseases associated with CD38 dysfunction include chronic lymphocytic leukemia and Richter's Syndrome.
Specifications
CD38 | |
Polyclonal | |
Unconjugated | |
CD38 | |
2'-phospho-ADP-ribosyl cyclase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-cyclic-ADP-ribose transferase; ADPRC 1; ADPRC1; ADP-ribosyl cyclase 1; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1; cADPr hydrolase 1; Cd38; CD38 antigen; CD38 antigen (ADP-ribosyl cyclase / cyclic ADP-ribose hydrolase); CD38 antigen (p45); CD38 antigen p45; CD38 molecule; CD38H; Cd38-rs1; cluster of differentiation 38; cyclic ADP-ribose hydrolase 1; ecto-nicotinamide adenine dinucleotide glycohydrolase; I-19; NAD(+) nucleosidase; NAD+ nucleosidase; NIM-R5 antigen; T10 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12494, 25668, 952 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.69 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
P28907, P56528, Q64244 | |
CD38 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 43-300 of human CD38 (NP_0017662). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.