Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD38 Rabbit anti-Human, Clone: 3, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP28965850UL
Description
CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
CD38 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
3 | |
Western Blot 1:2000-1:10000, ELISA 1:5000-1:10000, Immunoprecipitation 1-5 ul/mg of lysate | |
ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10 | |
Recombinant Human CD38 protein | |
50 μg | |
Apoptosis, B Cell Development and Differentiation Markers, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Stem Cell Markers | |
952 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction