Learn More
Description
The 90 monoclonal antibody specifically binds to CD38, a 42 kDa transmembrane glycoprotein on immature and mature, resting and activated, B lymphocytes. In contrast to humans, CD38 expression is down-regulated on mouse germinal center B cells and plasma cells. CD38 is also expressed on a subpopulation of thymic and peripheral T cells, NK cells, and splenic macrophages. Furthermore, CD38 has been detected on bone marrow-derived hematopoietic stem cells. The CD38 molecule is reported to exhibit both cyclase and hydrolase activities and plays a role in lymphocyte activation. CD31, both human and mouse, is reported to be a ligand for human CD38.
Specifications
Specifications
| Antigen | CD38 |
| Applications | Single Cell 3' Sequencing |
| Clone | 90/CD38 |
| Conjugate | Oligo |
| Host Species | Rat |
| Quantity | 25 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Mouse |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.