Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD39/ENTPD1 Antibody (498403), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4397X
Description
CD39/ENTPD1 Monoclonal antibody specifically detects CD39/ENTPD1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
CD39/ENTPD1 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
498403 | |
Flow Cytometry, CyTOF-ready | |
ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD39/ENTPD1, Thr38-Val478, Accession # P49961 | |
0.1 mL | |
Lipid and Metabolism, Myeloid derived Suppressor Cell | |
953 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction