Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD39/ENTPD1 Antibody (AC2), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250387AF700
Description
CD39/ENTPD1 Monoclonal specifically detects CD39/ENTPD1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
CD39/ENTPD1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ENTPD1 | |
EBV-transformed human B lymphoblastoid cell line | |
0.1 mL | |
Lipid and Metabolism, Myeloid derived Suppressor Cell | |
953 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry | |
AC2 | |
Western Blot, Flow Cytometry | |
ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction