Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD39 Monoclonal Antibody (24DMS1), Functional Grade, eBioscience™
GREENER_CHOICE

Rat Monoclonal Antibody

$431.65

Specifications

Antigen CD39
Clone 24DMS1
Concentration 1 mg/mL
Content And Storage 4°C
Applications Control, Flow Cytometry, Functional Assay, Western Blot
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-142-98
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
16039183
200 μg
$576.00
$431.65 / Each of 1

Save $144.35

25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Only null left
Add to Cart
 
Description

Description

Description: The 24DMS1 monoclonal antibody reacts with mouse CD39, also known as NTPDaseI. E-NTPDases are enzymes that convert nucleoside tri- and diphosphates (NTDPs) into nucleoside monophosphate (NMP), thereby removing toxic extracellular ATP and ADP. CD39 is the dominant member of this family in the immune system and is involved in suppression of inflammation and control of platelet activation. CD39 can impact expression of CD73, another E-NTPase. Together, these molecules influence inflammation responses. CD39 is expressed on B cells, Langerhans cells and most monocytes. In addition, CD39 is found on a subset of CD4+ T cells that are mostly CD25+FoxP3+ T reg cells. T reg cells from CD39-null mice showed impaired suppressive properties in vitro and in vivo. Applications Reported: This 24DMS1 antibody has been reported for use in flow cytometric analysis, and immunoblotting (WB). Applications Tested: This 24DMS1 antibody has been tested by flow cytometric analysis of mouse splenocytes and by WB. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. This antibody is provided as a functional grade but has not been tested in functional assay yet. Storage and...

CD39 is an integral membrane protein characterized by two transmembrane domains and forms a homotetramer structure. It is recognized as the most prominent ectoenzyme within the immune system, primarily functioning to remove toxic extracellular ATP by converting it into ADP or AMP. CD39 works in conjunction with CD73 to hydrolyze ATP and is well characterized on Langerhans cells. Originally identified on activated lymphocytes, CD39 is expressed on a variety of immune cells, including peripheral blood B cells, monocytes, and T cell clones. It is also found on a subset of T cells, B cells, and dendritic cells, with weaker expression observed on monocytes and granulocytes. The CD39 cell surface antigen is a 70-100kD molecule, and its expression is induced on T cells and increases on B cells as a late activation antigen. CD39 possesses intrinsic ecto-ATPase activity, playing a crucial role in modulating extracellular nucleotide levels and contributing to immune regulation.
Specifications

Specifications

CD39
1 mg/mL
Control, Flow Cytometry, Functional Assay, Western Blot
Functional Grade
Rat
RUO
PBS with no preservative; pH 7.2
P55772
12495
Primary
Affinity chromatography
24DMS1
4°C
Monoclonal
Liquid
IgG2b κ
Mouse
ENTPD1
2610206B08Rik; AA408691; ATPDase; Cd39; CD39 antigen; DKFZp686D194; DKFZp686I093; ecto-apyrase; Ecto-ATP diphosphohydrolase 1; ecto-ATPase 1; ecto-ATPDase 1; Ectonucleoside triphosphate diphosphohydrolase 1; ENTPD1; FLJ40921; FLJ40959; Lymphoid cell activation antigen; NTPDase 1; NTPDase-1; SPG64
ENTPD1
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.