Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD39 Monoclonal Antibody (eBioA1 (A1)), NovaFluor™ Red 725, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD39 |
---|---|
Clone | eBioA1 (A1) |
Concentration | 0.6 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H032T03R05A
![]() |
Invitrogen™
H032T03R05A |
100 Tests |
Each for $430.00
|
|
|||||
H032T02R05A
![]() |
Invitrogen™
H032T02R05A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD39 Monoclonal antibody specifically detects CD39 in Human samples. It is validated for Flow CytometrySpecifications
CD39 | |
0.6 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P49961 | |
953 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
eBioA1 (A1) | |
Flow Cytometry | |
NovaFluor Red 725 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
2610206B08Rik; AA408691; ATPDase; Cd39; CD39 antigen; DKFZp686D194; DKFZp686I093; ecto-apyrase; Ecto-ATP diphosphohydrolase 1; ecto-ATPase 1; ecto-ATPDase 1; Ectonucleoside triphosphate diphosphohydrolase 1; ENTPD1; FLJ40921; FLJ40959; Lymphoid cell activation antigen; NTPDase 1; NTPDase-1; SPG64 | |
IgG1 κ | |
Affinity chromatography | |
ENTPD1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title