Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD39 Mouse anti-Human, Brilliant Violet 605, Clone: TU66 (also known as Tü 66, Tü66), BD Optibuild™

Catalog No. BDB742522 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742522 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742522 Supplier BD Biosciences Supplier No. 742522
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TU66 monoclonal antibody reacts with human CD39, also known as ectonucleoside triphosphate diphosphohydrolase 1 (ENTPD1), an ectoenzyme that degrades ATP to AMP. It is a member of the ectonucleoside triphosphate dihydrolases (E-NTPDases) family involved in the regulation of extracellular nucleotide catabolism by controlling the extracellular nucleoside triphosphate pool (NTP). CD39 is expressed on a subset of T cells, B cells and dendritic cells, with weak staining of monocytes and granulocytes. Recently, CD39 has been found to be expressed primarily by immune-suppressive Foxp3(+) regulatory T (Treg) cells in both human and mice. In humans, CD39 is restricted to a subset of Foxp3+ regulatory effector/memory-like T cells. In mice, the enzyme is present on most, if not all, CD4+CD25+ cells. CD39 expression is driven by Foxp3 and it is thought that CD39 allows Treg cells to enter inflamed areas where high levels of ATP are present.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD39
Classification Monoclonal
Clone TU66 (also known as Tü 66, Tü66)
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ENTPD1, NTPDase-1, Ecto-ATPase 1, Ecto-ATPDase 1
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.