Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD39 Mouse anti-Human, Oligo, Clone: TU66 (also known as Tü 66, Tü66), BD™ AbSeq ™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 940073
This item is not returnable.
View return policy
Description
CD39 Monoclonal antibody specifically detects CD39 in Human samples. It is validated for Single Cell 3' Sequencing
Specifications
CD39 | |
Monoclonal | |
Oligo | |
ENTPD1; NTPDase-1; Ecto-ATPase 1; Ecto-ATPDase 1 | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Single Cell 3' Sequencing | |
Clone TU66 (also known as Tü 66, Tü66) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction