Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD39 Mouse anti-Human, R718, Clone: A1 (also known as A1/CD39), BD Horizon™
Mouse Monoclonal Antibody
$164.00 - $373.50
Specifications
Antigen | CD39 |
---|---|
Clone | A1 (also known as A1/CD39) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Description
CD39 Monoclonal antibody specifically detects CD39 in Human samples. It is validated for Flow CytometrySpecifications
CD39 | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
A1; ENTPD1; NTPDase-1; ecto-ATPDase 1; ecto-ATPase 1; ecto-apyrase | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |
A1 (also known as A1/CD39) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
PHA-activated Human Peripheral Blood Mononuclear Cells | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title