Learn More
Invitrogen™ CD39 Polyclonal Antibody

Description
Positive test controls include: U-251MG, HeLa, A-549, LO2, HL-60, MCF7, Mouse lung, Mouse spleen, Rat kidney. The target is usually found in the following locations: Membrane, Multi-pass membrane protein. Immunogen sequence: MESEELADRV LDVVERSLSN YPFDFQGARI ITGQEEGAYG WITINYLLGK FSQKTRWFSI VPYETNNQET FGALDLGGAS TQVTFVPQNQ TIESPDNALQ FRLYGKDYNV YTHSFLCYGK DQALWQKLAK DIQVASNEIL RDPCFHPGYK KVVNVSDLYK TPCTKRFEMT LPFQQFEIQG.
Specifications
Specifications
| Antigen | CD39 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | ENTPD1 |
| Gene Accession No. | P49961, P55772, P97687 |
| Gene Alias | 2610206B08Rik; AA408691; ATPDase; Cd39; CD39 antigen; DKFZp686D194; DKFZp686I093; ecto-apyrase; Ecto-ATP diphosphohydrolase 1; ecto-ATPase 1; ecto-ATPDase 1; Ectonucleoside triphosphate diphosphohydrolase 1; ENTPD1; FLJ40921; FLJ40959; Lymphoid cell activation antigen; NTPDase 1; NTPDase-1; SPG64 |
| Gene Symbols | ENTPD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.