Learn More
Description
Specifications
Specifications
| Antigen | CD39 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Y23-1185 |
| Conjugate | Brilliant Violet 421 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | ectonucleoside triphosphate diphosphohydrolase 1; Entpd1; ecto-ATP diphosphohydrolase 1; ecto-ATPDase 1; ecto-ATPase 1; Cd39; NTPDase 1 |
| Host Species | Rat |
| Immunogen | Mouse CD39 Transfected Cell Line |
| Quantity | 50 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.