Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD39L3/ENTPD3 Antibody (101), DyLight 755 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289707IR
Description
CD39L3/ENTPD3 Monoclonal antibody specifically detects CD39L3/ENTPD3 in Human samples. It is validated for ELISASpecifications
CD39L3/ENTPD3 | |
Monoclonal | |
DyLight 755 | |
CD39 antigen-like 3, CD39L3EC 3.6.1.5, CD39-like 3, EC 3.6.1, Ecto-apyrase 3, Ecto-ATP diphosphohydrolase 3, Ecto-ATPase 3, Ecto-ATPDase 3, ectonucleoside triphosphate diphosphohydrolase 3, FLJ93839, HB6NTPDase 3, NTPDase-3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD39L3/ENTPD3 (O75355; Gln44-Pro485). | |
0.1 mL | |
Lipid and Metabolism | |
956 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
101 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction