Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD39L3/ENTPD3 Rat anti-Mouse, Alexa Fluor 647, Clone: 568610, R&D Systems™

Catalog No. FAB4464R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4464R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4464R100 Supplier R&D Systems Supplier No. FAB4464R100UG
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD39L3/ENTPD3 Monoclonal antibody specifically detects CD39L3/ENTPD3 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD39L3/ENTPD3
Applications Flow Cytometry
Classification Monoclonal
Clone 568610
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_848791
Gene Alias ATPDase, CD39 antigen-like 3, CD39L3, CD39L3EC 3.6.1.5, CD39-like 3, EC 3.6.1, Ecto-apyrase 3, Ecto-ATP diphosphohydrolase 3, Ecto-ATPase 3, Ecto-ATPDase 3, ectonucleoside triphosphate diphosphohydrolase 3, ENTPD3, FLJ93839, HB6, HB6NTPDase 3, NTPDase-3
Gene Symbols ENTPD3
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CD39L3/ENTPD3 Gln44-Pro485 Accession # NP_848791
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 956
Test Specificity Detects mouse CD39L3/ENTPD3 in direct ELISAs. In Western blots, less than 15% cross-reactivity with recombinant human (rh) CD39L4 and no cross-reactivity with rhCD39L3, recombinant mouse (rm) CD39, or rmCD39L2 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.