Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD39L4/ENTPD5 Antibody (SR1805), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322057100UL
Description
CD39L4/ENTPD5 Monoclonal antibody specifically detects CD39L4/ENTPD5 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CD39L4/ENTPD5 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1805 | |
Western Blot 1:500-1:2000 | |
CD39 antigen-like 4, CD39L4NTPDase 5, CD39-like 4, EC 3.6.1, EC 3.6.1.42, EC 3.6.1.6, ectonucleoside triphosphate diphosphohydrolase 5, ER-UDPase, GDPase ENTPD5, Guanosine-diphosphatase ENTPD5, MGC163357, MGC163359, NTPDase-5, Nucleoside diphosphatase, PCPH, Pcph proto-oncogene protein, proto-oncogene CPH, UDPase ENTPD5, Uridine-diphosphatase ENTPD5 | |
A synthesized peptide derived from human CD39L4/ENTPD5 (Uniprot #: O75356) | |
100 μg | |
Lipid and Metabolism | |
957 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction