Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd3e, anti-Mouse, Clone: 145-2C11, Abnova™

Catalog No. 89032421 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-421 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-421 Supplier Abnova Corporation Supplier No. RAB00078
Add to Cart
Add to Cart

Monoclonal Antibody

Specifications

Antigen Cd3e
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 145-2C11
Conjugate Unconjugated
Dilution Flow Cytometry (1 ug/mL) Immunoprecipitation (10 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% Proclin 300).
Gene CD3 antigen, epsilon polypeptide
Gene Alias AI504783/CD3/CD3epsilon/T3e
Gene Symbols Cd3e
Immunogen Original antibody is raised against H-2Kb-specific murine cytotoxic T-lymphocyte (CTL) clone.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12501
Test Specificity This antibody recognizes a 25 kDa protein component of the antigen-specific T-cell receptor.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.