Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD3e/CD8 Antibody Cocktail, FITC, PE
GREENER_CHOICE

Catalog No. PIMA516606
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516606 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516606 Supplier Invitrogen™ Supplier No. MA516606
Add to Cart
Add to Cart

Antibody Cocktail

This antibody recognizes canine CD3 epsilon and CD8 cell surface antigens. Clone CA17.2A12 is a FITC-conjugated mouse IgG1 that recognizes canine CD3, a T cell differentiation antigen expressed on mature T lymphocytes. Clone YCATE 55.9 is an RPE-conjugated Rat IgG1 that recognizes canine CD8 epsilon, which is expressed by cytotoxic T cells. YCATE 55.9 binds to CD8 alpha. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

putative component of the CD3 T-cell receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3e/CD8
Applications Flow Cytometry
Classification Cocktail
Clone CA17.2A12/YCATE55.9
Concentration Conc. Not Determined
Conjugate FITC/PE
Formulation PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
Gene CD8A
Gene Accession No. P27597, P33706
Gene Alias AI504783; BB154331; CD3; CD3 antigen delta polypeptide; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 delta; CD3 epsilon; CD3 gamma-chain; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; CD3d; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; Cd3e; CD3e antigen, epsilon polypeptide (TiT3 complex); CD3e molecule; CD3e molecule, epsilon (CD3-TCR complex); CD3epsilon; CD3-epsilon; Cd3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; CD8; CD8 antigen; CD8 antigen 32 kDa chain; CD8 antigen 37 kDa chain; CD8 antigen alpha-chain; CD8 antigen beta-chain; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide (p32); CD8 antigen, alpha-chain; CD8 antigen, beta chain; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8A; CD8a molecule; CD8alpha; CD8B; CD8b molecule; CD8B1; CD8beta; Ctg3; Ctg-3; FLJ18683; IMD17; IMD18; IMD19; LEU2; Leu-2; Leu2 T-lymphocyte antigen; Ly-2; LY3; Ly-35; Ly-B; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at 100); LYT3; MAL; OKT3, delta chain; OKT8 T-cell antigen; OX-8 membrane antigen; p32; P37; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T3D; T3E; T3G; T8 T-cell antigen; T-cell antigen Leu2; T-cell antigen receptor complex, epsilon subunit of T3; T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-2; TCRE; T-lymphocyte differentiation antigen T8/Leu-2
Gene Symbols CD3D, CD3E, CD3G, CD8A, Cd8b
Host Species Mixed
Immunogen Affinity enriched TCR/CD3 membrane proteins isolated from thymocytes and the T cell line CLGL-9, Canine CD8 alpha chimaeric human IgG1 Fc fusion protein.
Purification Method Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403157, 442981, 479419, 483076, 489383
Target Species Canine
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.