Learn More
CD3e Hamster anti-Mouse, PE-Cy7, Clone: 145-2C11, BD
Hamster Monoclonal Antibody
$104.00 - $246.00
Specifications
Antigen | CD3e |
---|---|
Clone | 145-2C11 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The 145-2C11 monoclonal antibody specifically binds to the 25-kDa ε chain of the T-cell receptor-associated CD3 complex that is expressed on thymocytes, mature T lymphocytes, and NK-T cells. The cytoplasmic domain of CD3e participates in the signal transduction events that activate several cellular biochemical pathways as a result of antigen recognition. Soluble 145-2C11 antibody can activate either unprimed (naive) or primed (memory/preactivated) T cells in vivo or in vitro, in the presence of Fc receptor-bearing accessory cells. In contrast, plate-bound 145-2C11 can activate T cells in the absence of accessory cells. Soluble 145-2C11 antibody has been reported to induce re-directed lysis of Fc receptor-bearing target cells by CTL clones and can also block lysis of specific target cells by antigen-specific CTL's. Under some conditions, T-cell activation by 145-2C11 antibody has been reported to result in apoptotic cell death. The 145-2C11 antibody does not cross-react with rat leukocytes. Preincubation of thymus cell suspensions at 37°C for 2-4 hours prior to staining reportedly enhances the ability of anti-CD3ε and anti-αβ TCR mAbs to detect the T-cell receptor on immature thymocytes.
Specifications
CD3e | |
0.2mg/mL | |
Monoclonal | |
Hamster | |
Mouse | |
H-2Kb specific cytotoxic T lymphocyte clone BM10-37 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
145-2C11 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
CD3; CD3 epsilon; Cd3e; CD3ε; T3e |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.