Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3E Rabbit anti-Human, Unconjugated, Clone: C3e/3171R, Abnova™

Catalog No. 89339680 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-680 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-680 Supplier Abnova Corporation Supplier No. RAB00665
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CD3E Monoclonal antibody specifically detects CD3E in Human samples. It is validated for ELISA, IHC-P.

Specifications

Antigen CD3E
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C3e/3171R
Conjugate Unconjugated
Description CD3e molecule, epsilon (CD3-TCR complex)
Gene CD3E
Gene Alias FLJ18683, T3E, TCRE
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 23-119 of human CD3E
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 916
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.