Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD3g Recombinant Rabbit Monoclonal Antibody (7J2L5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542846
This item is not returnable.
View return policy
Description
Positive test controls include: Rat lung, Rat spleen. Immunogen sequence: GMYQCKGSQN KSKPLQVYYR MCQNCIELNA ATISGFLFAE IVSIFVLAVG VYFIAGQDGV RQSRASDKQT LLPNDQLYQP LKDREDDQYS HLQGNQLRRN.
CD3G is the CD3-gamma polypeptide, which together with CD3-epsilon, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. Defects in the CD3G gene are associated with T cell immunodeficiency.
Specifications
CD3g | |
Recombinant Monoclonal | |
0.78 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09693, Q64159 | |
CD3G | |
A synthetic peptide corresponding to a sequence within amino acids 83-182 of human CD3G (P09693). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
7J2L5 | |
Unconjugated | |
CD3G | |
CD3 antigen, gamma polypeptide; CD3 gamma-chain; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; Cd3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; Ctg3; Ctg-3; IMD17; T3G; T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 gamma chain; T-cell surface glycoprotein CD3 gamma chain | |
Rabbit | |
Affinity chromatography | |
RUO | |
300678, 917 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction