Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (AV309A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260889
Description
CD4 Monoclonal specifically detects CD4 in Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD4 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Mouse | |
Unpurified | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
920 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
AV309A | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
CD4 | |
Bovine mononuclear leukocytes | |
0.1 mg | |
Primary | |
Chicken | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction