Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (C4/206), Novus Biologicals™

Mouse Monoclonal Antibody
$280.00 - $524.00
Specifications
Antigen | CD4 |
---|---|
Clone | C4/206 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, SDS-Page, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24532900
![]() |
Novus Biologicals
NBP2453290.02MG |
0.02 mg |
Each for $280.00
|
|
|||||
NBP24532901
![]() |
Novus Biologicals
NBP2453290.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP24532902
![]() |
Novus Biologicals
NBP2453290.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for ELISA.Specifications
CD4 | |
0.2 mg/mL | |
Flow Cytometry, SDS-Page, Immunofluorescence | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Recombinant human CD4 protein | |
Primary | |
Store at 4C. | |
55 kDa |
C4/206 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P01730 | |
920 | |
IgG1 κ | |
Protein A or G purified | |
Recognizes a protein of 55kDa, identified as CD4. CD4 is a membrane glycoprotein of T lymphocytes that interacts with major histocompatibility complex class II antigens and is also a receptor for the human immunodeficiency virus. This protein is expressed not only in T lymphocytes, but also in B cells, macrophages, and granulocytes. It is also expressed in specific regions of the brain. The protein functions to initiate or augment the early phase of T-cell activation, and may function as an important mediator of indirect neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcript variants encoding different isoforms have been identified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title