Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (CE9.1 (Clenoliximab)), Alexa Fluor™ 405, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275910AF405
Description
CD4 Monoclonal specifically detects CD4 in Human, Chimpanzee samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.Specifications
CD4 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD4 | |
Domain 1 of human CD4. | |
0.1 mL | |
Primary | |
The antibody binds to human CD4 with a Kd of 1.0 nM and an ED50 of 0.2 ug/ml. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
CE9.1 (Clenoliximab) | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Human | |
Protein A purified | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
920 | |
Human, Primate | |
IgG4 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction