Learn More
CD4 Antibody (CE9.1 (Clenoliximab)), Alexa Fluor™ 594 - Chimeric, Novus Biologicals™
SDP
Rabbit Monoclonal Antibody
Supplier:  Novus Biologicals NBP252681AF594
Catalog No. NP22681A94
Only null left
Add to Cart
Description
Description
CD4 Monoclonal specifically detects CD4 in Human, Chimpanzee samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.
Specifications
Specifications
CD4
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
CD4
Domain 1 of human CD4 Antibody
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers
920
Human, Primate
IgG λ
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
CE9.1 (Clenoliximab)
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rabbit
0.1 mL
Primary
The antibody binds to human CD4 with a Kd of 1.0 nM and an ED50 of 0.2 ug/ml.
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.