Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (CE9.1 (Clenoliximab)), PerCP, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275910PCP
Description
CD4 Monoclonal specifically detects CD4 in Human, Chimpanzee samples. It is validated for Flow Cytometry.Specifications
| CD4 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| CD4 | |
| Domain 1 of human CD4. | |
| 0.1 mL | |
| Primary | |
| The antibody binds to human CD4 with a Kd of 1.0 nM and an ED50 of 0.2 ug/ml. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| CE9.1 (Clenoliximab) | |
| Flow Cytometry | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Human | |
| Protein A purified | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| 920.0 | |
| Human, Primate | |
| IgG4 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction