Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (GK1.5) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$501.50
Specifications
Antigen | CD4 |
---|---|
Clone | GK1.5 |
Concentration | 0.5mg/mL |
Dilution | Flow Cytometry 2.5 ug 1 million cells, Immunohistochemistry 1-25 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface) 2.5 ug 1 million cells, Cell depletion, Inhibition Activity, CyTOF-ready |
Classification | Monoclonal |
Description
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface), Cell depletion, Inhibition Activity, CyTOF-ready.Specifications
CD4 | |
0.5mg/mL | |
Monoclonal | |
Purified | |
RUO | |
NP_000607 | |
920 | |
The mouse cytotoxic T lymphocyte line V4 was used as the immunogen for the CD4 antibody. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
GK1.5 | |
Flow Cytometry 2.5 ug 1 million cells, Immunohistochemistry 1-25 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface) 2.5 ug 1 million cells, Cell depletion, Inhibition Activity, CyTOF-ready | |
Unconjugated | |
Rat | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
CD4 | |
IgG κ | |
Protein G purified | |
This product reacts with Mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title