Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (MT310) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$280.00
Specifications
Antigen | CD4 |
---|---|
Clone | MT310 |
Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25266325
![]() |
Novus Biologicals
NBP2526630.025MG |
0.025 mg |
Each of 1 for $280.00
|
|
|||||
Description
CD4 Monoclonal specifically detects CD4 in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunofluorescence, CyTOF-ready.Specifications
CD4 | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
CD4 | |
IgG | |
Protein A or G purified |
MT310 | |
Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
920 | |
CD4 | |
Primary | |
mt310 binds to the extracellular domain D1 of CD4 - the same site as the HIV-1 gp120-binding site |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title