Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | CD4 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-3 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056457T
![]() |
Novus Biologicals
NB10056457SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056457
![]() |
Novus Biologicals
NB10056457 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
CD4 Polyclonal specifically detects CD4 in Human, Primate samples. It is validated for Western Blot.Specifications
CD4 | |
Western Blot 1-3 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Primate | |
NP_000607 | |
920 | |
A portion of amino acids 325-375 of human CD4 Antibody was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
CD4 | |
IgG | |
Protein G purified | |
The amino acid sequence used as immunogen is 100% homologous in human and chimp, and 93% homologous in monkey. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title